missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-methylthiazole 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC392790250

 View more versions of this product

Catalog No. AC392790250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Amino-5-methylthiazole
Authentic
100.0
Brown
≥97.5%
98%
114.17
7305-71-7
97.5
Glass bottle
Powder
25g
C4H6N2S
MFCD00078317

Chemical Identifiers

7305-71-7
100.0
C4H6N2S
MFCD00078317
≥97.5%
97.5
98%
114.17
2-Amino-5-methylthiazole

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning