Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-methylthiazole, 98+%

Chemical Identifiers

Chemical Name or Material 2-Amino-5-methylthiazole
CAS 7305-71-7
Molecular Formula C4H6N2S
Formula Weight 114.17
MDL Number MFCD00078317
UN Number UN3077
Beilstein 109603
Synonym 5-Methyl-2-thiazolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1103906

  SDS
alfa aesar™
L1103906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.17
Chemical Name or Material 2-Amino-5-methylthiazole
Health Hazard 2 GHS H Statement
H301-H373
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P264-P301+P310-P321-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
MDL Number MFCD00078317
Molecular Formula C4H6N2S
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Synonym 5-Methyl-2-thiazolamine
Odor Recognizable
UN Number UN3077
Melting Point 95° to 98°C
Quantity 5g
CAS 7305-71-7
Beilstein 109603
Safety and Handling

Danger

GHS P Statement
P260-P264-P301+P310-P321-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H373
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.