Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-methylpyridine-3-carboxaldehyde, 95%

Chemical Identifiers

Chemical Name or Material 2-Amino-5-methylpyridine-3-carboxaldehyde
CAS 1023814-35-8
Formula Weight 136.15
MDL Number MFCD09754122
Beilstein 23782189
Synonym 2-Amino-5-methylnicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66175MD

 
alfa aesar™
H66175MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.15
CAS 1023814-35-8
Chemical Name or Material 2-Amino-5-methylpyridine-3-carboxaldehyde
MDL Number MFCD09754122
Assay 95%
Physical Form Solid
Quantity 250mg
Synonym 2-Amino-5-methylnicotinaldehyde
Beilstein 23782189