Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-methylhexane, 98%

Chemical Identifiers

Chemical Name or Material 2-Amino-5-methylhexane
CAS 28292-43-5
Molecular Formula C7H17N
Formula Weight 115.22
MDL Number MFCD00014813
UN Number UN2733
Beilstein 1731716
Synonym 1,4-Dimethylpentylamine; 5-Methyl-2-hexylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0863809

  SDS
alfa aesar™
L0863809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0863818

  SDS
alfa aesar™
L0863818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 115.22
Chemical Name or Material 2-Amino-5-methylhexane
MDL Number MFCD00014813
Assay 98%
CAS 28292-43-5
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
TSCA TSCA
Synonym 1,4-Dimethylpentylamine; 5-Methyl-2-hexylamine
Boiling Point 128° to 129°C
Molecular Formula C7H17N
Flash Point 28°C (82°F)
UN Number UN2733
Quantity 10g
RTECS Number SC0875000
Beilstein 1731716
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.