missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-methylbenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC160200250

 View more versions of this product

Catalog No. AC160200250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-5-methylbenzoic acid
Plastic bottle
174.0°C to 177.0°C
2941-78-8
100.0
C8H9NO2
MFCD00007909
2-amino-5-methyl benzoic acid
CC1=CC(=C(C=C1)N)C(=O)O
151.16
151.16
97%
Authentic
White to Beige
25g
96.0
96% min. (HPLC)
H2NC6H3(CH3)CO2H
14, 481
NBUUUJWWOARGNW-UHFFFAOYSA-N
2-amino-5-methylbenzoic acid
76255
Powder

Chemical Identifiers

2941-78-8
151.16
NBUUUJWWOARGNW-UHFFFAOYSA-N
76255
CC1=CC(=C(C=C1)N)C(=O)O
C8H9NO2
MFCD00007909
2-amino-5-methyl benzoic acid
2-amino-5-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-932-3

SDS Documents