missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-methoxybenzonitrile, 95%, ACROS Organics™

Chemical Identifiers

CAS 23842-82-2
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.16
InChI Key SRWMPAZUWXLIPG-UHFFFAOYSA-N
Synonym 2-amino-5-methoxy-benzonitrile, 2-cyano-4-methoxyaniline, 4-amino-3-cyanoanisole, benzonitrile, 2-amino-5-methoxy, benzonitrile,2-amino-5-methoxy, ksc495g0f
PubChem CID 3513002
IUPAC Name 2-amino-5-methoxybenzonitrile
SMILES COC1=CC(=C(C=C1)N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451340010
SDS View Documents
Thermo Fisher Scientific
AC451340010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451340050
SDS View Documents
Thermo Fisher Scientific
AC451340050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

23842-82-2
148.16
2-amino-5-methoxy-benzonitrile, 2-cyano-4-methoxyaniline, 4-amino-3-cyanoanisole, benzonitrile, 2-amino-5-methoxy, benzonitrile,2-amino-5-methoxy, ksc495g0f
2-amino-5-methoxybenzonitrile
C8H8N2O
SRWMPAZUWXLIPG-UHFFFAOYSA-N
3513002
COC1=CC(=C(C=C1)N)C#N

Specifications

1g
94% min. (GC)
2-amino-5-methoxy-benzonitrile, 2-cyano-4-methoxyaniline, 4-amino-3-cyanoanisole, benzonitrile, 2-amino-5-methoxy, benzonitrile,2-amino-5-methoxy, ksc495g0f
COC1=CC(=C(C=C1)N)C#N
148.16
148.16
2-Amino-5-methoxybenzonitrile
23842-82-2
C8H8N2O
SRWMPAZUWXLIPG-UHFFFAOYSA-N
2-amino-5-methoxybenzonitrile
3513002
95%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS