missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-methoxybenzonitrile, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451340050
Specifications
2-Amino-5-methoxybenzonitrile | |
23842-82-2 | |
C8H8N2O | |
SRWMPAZUWXLIPG-UHFFFAOYSA-N | |
2-amino-5-methoxybenzonitrile | |
3513002 | |
95% |
5g | |
94% min. (GC) | |
2-amino-5-methoxy-benzonitrile, 2-cyano-4-methoxyaniline, 4-amino-3-cyanoanisole, benzonitrile, 2-amino-5-methoxy, benzonitrile,2-amino-5-methoxy, ksc495g0f | |
COC1=CC(=C(C=C1)N)C#N | |
148.16 | |
148.16 |
Chemical Identifiers
23842-82-2 | |
148.16 | |
2-amino-5-methoxy-benzonitrile, 2-cyano-4-methoxyaniline, 4-amino-3-cyanoanisole, benzonitrile, 2-amino-5-methoxy, benzonitrile,2-amino-5-methoxy, ksc495g0f | |
2-amino-5-methoxybenzonitrile |
C8H8N2O | |
SRWMPAZUWXLIPG-UHFFFAOYSA-N | |
3513002 | |
COC1=CC(=C(C=C1)N)C#N |
Safety and Handling
GHS Signal Word: Warning