missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-mercapto-1,3,4-thiadiazole, 98+%

Manufacturer:  Alfa Aesar™ A1145114

 View more versions of this product

Catalog No. AAA1145114



Specifications

Specifications

2-Amino-5-mercapto-1,3,4-thiadiazole
≥98%
~243°C (decomposition)
81728
5-Amino-1,3,4-thiadiazole-2-thiol
C1(=NNC(=S)S1)N
133.187
133.19
2349-67-9
C2H3N3S2
MFCD00003108
25g
GDGIVSREGUOIJZ-UHFFFAOYSA-N
5-amino-3H-1,3,4-thiadiazole-2-thione
2723847

Chemical Identifiers

2349-67-9
133.187
GDGIVSREGUOIJZ-UHFFFAOYSA-N
2723847
C1(=NNC(=S)S1)N
C2H3N3S2
MFCD00003108
5-Amino-1,3,4-thiadiazole-2-thiol
5-amino-3H-1,3,4-thiadiazole-2-thione
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI4550000

TSCA : TSCA

SDS Documents