Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-iodopyridine-3-methanol, 97%

$247.82 - $1,097.50

Chemical Identifiers

Chemical Name or Material 2-Amino-5-iodopyridine-3-methanol
CAS 618107-90-7
Molecular Formula C6H7IN2O
Formula Weight 250.04
MDL Number MFCD05663524
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5001103

 
alfa aesar™
H5001103
1g Each for $247.82

AAH5001106

 
alfa aesar™
H5001106
5g Each for $1,097.50
Description & Specifications

Specifications

Formula Weight 250.04
Chemical Name or Material 2-Amino-5-iodopyridine-3-methanol
Assay 97%
Molecular Formula C6H7IN2O
CAS 618107-90-7
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD05663524
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.