Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-iodo-4-methylpyridine, 95%

$126.72 - $287.80

Chemical Identifiers

Chemical Name or Material 2-Amino-5-iodo-4-methylpyridine
CAS 356561-08-5
Molecular Formula C6H7IN2
Formula Weight 234.04
MDL Number MFCD06637727
Synonym 2-Amino-5-iodo-4-picoline; 5-Iodo-4-methyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2708903

  SDS
alfa aesar™
H2708903
1g Each for $287.80

AAH27089MD

  SDS
alfa aesar™
H27089MD
250mg Each for $126.72
Description & Specifications

Specifications

Formula Weight 234.04
MDL Number MFCD06637727
Synonym 2-Amino-5-iodo-4-picoline; 5-Iodo-4-methyl-2-pyridinamine
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
Chemical Name or Material 2-Amino-5-iodo-4-methylpyridine
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
CAS 356561-08-5
Molecular Formula C6H7IN2
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.