Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-iodo-4,6-dimethylpyridine, 95%

$120.38 - $270.92

Chemical Identifiers

Chemical Name or Material 2-Amino-5-iodo-4,6-dimethylpyridine
CAS 885952-12-5
Molecular Formula C7H9IN2
Formula Weight 248.07
MDL Number MFCD06798226
Synonym 4,6-Dimethyl-5-iodo-2-pyridinamine; 6-Amino-2,4-dimethyl-3-iodopyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2720203

 
alfa aesar™
H2720203
1g Each for $270.92

AAH27202MD

 
alfa aesar™
H27202MD
250mg Each for $120.38
Description & Specifications

Specifications

Formula Weight 248.07
Chemical Name or Material 2-Amino-5-iodo-4,6-dimethylpyridine
Melting Point 164° to 166°C
Assay 95%
Quantity 1g
Molecular Formula C7H9IN2
MDL Number MFCD06798226
CAS 885952-12-5
Synonym 4,6-Dimethyl-5-iodo-2-pyridinamine; 6-Amino-2,4-dimethyl-3-iodopyridine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.