Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-iodo-3-methylpyridine, 95%

$94.93 - $215.47

Chemical Identifiers

Chemical Name or Material 2-Amino-5-iodo-3-methylpyridine
CAS 166266-19-9
Molecular Formula C6H7IN2
Formula Weight 234.04
MDL Number MFCD02102422
Synonym 2-Amino-5-iodo-3-picoline; 5-Iodo-3-methyl-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2751003

  SDS
alfa aesar™
H2751003
1g Each for $215.47

AAH27510MD

  SDS
alfa aesar™
H27510MD
250mg Each for $94.93
Description & Specifications

Specifications

Formula Weight 234.04
Health Hazard 1 Danger
Quantity 1g
Chemical Name or Material 2-Amino-5-iodo-3-methylpyridine
Assay 95%
Synonym 2-Amino-5-iodo-3-picoline; 5-Iodo-3-methyl-2-pyridinamine
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 166266-19-9
MDL Number MFCD02102422
Molecular Formula C6H7IN2
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.