Learn More
2-Amino-5-hydroxypyrimidine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 434330010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Amino-5-hydroxypyrimidine | |
94.0 | |
Authentic | |
Glass bottle | |
MFCD08062411 | |
2-amino-5-hydroxypyrimidine, 5-pyrimidinol, 2-amino, 2-amino-pyrimidin-5-ol, 5-hydroxypyrimidin-2-amine, 2-amino-5-hydroxy pyrimidine, 2-amino-5-pyrimidinol, zlchem 1241, pubchem21503, 5-hydroxy-2-aminopyrimidine, acmc-1c7b8 | |
NC1=NC=C(O)C=N1 | |
111.10 | |
111.1 |
143489-45-6 | |
100.0 | |
94% min. (HPLC) | |
C4H5N3O | |
1g | |
NVYMOVCPYONOSF-UHFFFAOYSA-N | |
2-aminopyrimidin-5-ol | |
17750388 | |
95% |
Chemical Identifiers
143489-45-6 | |
111.10 | |
NVYMOVCPYONOSF-UHFFFAOYSA-N | |
17750388 | |
NC1=NC=C(O)C=N1 |
C4H5N3O | |
MFCD08062411 | |
2-amino-5-hydroxypyrimidine, 5-pyrimidinol, 2-amino, 2-amino-pyrimidin-5-ol, 5-hydroxypyrimidin-2-amine, 2-amino-5-hydroxy pyrimidine, 2-amino-5-pyrimidinol, zlchem 1241, pubchem21503, 5-hydroxy-2-aminopyrimidine, acmc-1c7b8 | |
2-aminopyrimidin-5-ol |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only