missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-hydroxypyrimidine, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC434330050

 View more versions of this product

Catalog No. AC434330050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-5-hydroxypyrimidine
Glass bottle
5g
94.0
95%
2-amino-5-hydroxypyrimidine, 5-pyrimidinol, 2-amino, 2-amino-pyrimidin-5-ol, 5-hydroxypyrimidin-2-amine, 2-amino-5-hydroxy pyrimidine, 2-amino-5-pyrimidinol, zlchem 1241, pubchem21503, 5-hydroxy-2-aminopyrimidine, acmc-1c7b8
C1=C(C=NC(=N1)N)O
111.104
111.1
≥94.0%
Authentic
White to Yellow
143489-45-6
100.0
C4H5N3O
NVYMOVCPYONOSF-UHFFFAOYSA-N
2-aminopyrimidin-5-ol
17750388
Powder or Crystalline Powder

Chemical Identifiers

143489-45-6
111.104
2-amino-5-hydroxypyrimidine, 5-pyrimidinol, 2-amino, 2-amino-pyrimidin-5-ol, 5-hydroxypyrimidin-2-amine, 2-amino-5-hydroxy pyrimidine, 2-amino-5-pyrimidinol, zlchem 1241, pubchem21503, 5-hydroxy-2-aminopyrimidine, acmc-1c7b8
2-aminopyrimidin-5-ol
C4H5N3O
NVYMOVCPYONOSF-UHFFFAOYSA-N
17750388
C1=C(C=NC(=N1)N)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents