Learn More
2-Amino-5-hydroxybenzoic acid, 98%, Thermo Scientific Chemicals
$201.08 - $201.08
Chemical Identifiers
CAS | 394-31-0 |
---|---|
Molecular Formula | C7H7NO3 |
Molecular Weight (g/mol) | 153.137 |
MDL Number | MFCD00007870 |
InChI Key | HYNQTSZBTIOFKH-UHFFFAOYSA-N |
Synonym | 5-hydroxyanthranilic acid, diabeton, benzoic acid, 2-amino-5-hydroxy, 2-amino-5-hydroxy benzoic acid, unii-uf9ww3i410, 2-amino-5-hydroxy-benzoic acid, 5-hydroxyanthranillicacid, pubchem15307, 4-amino-3-carboxyphenol, 5-hydroxyanthranillic acid |
PubChem CID | 164592 |
IUPAC Name | 2-amino-5-hydroxybenzoic acid |
SMILES | C1=CC(=C(C=C1O)C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0825614
|
Thermo Scientific Chemicals
L0825614 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
394-31-0 | |
153.137 | |
HYNQTSZBTIOFKH-UHFFFAOYSA-N | |
164592 | |
C1=CC(=C(C=C1O)C(=O)O)N |
C7H7NO3 | |
MFCD00007870 | |
5-hydroxyanthranilic acid, diabeton, benzoic acid, 2-amino-5-hydroxy, 2-amino-5-hydroxy benzoic acid, unii-uf9ww3i410, 2-amino-5-hydroxy-benzoic acid, 5-hydroxyanthranillicacid, pubchem15307, 4-amino-3-carboxyphenol, 5-hydroxyanthranillic acid | |
2-amino-5-hydroxybenzoic acid |
Specifications
394-31-0 | |
C7H7NO3 | |
25 g | |
5-hydroxyanthranilic acid, diabeton, benzoic acid, 2-amino-5-hydroxy, 2-amino-5-hydroxy benzoic acid, unii-uf9ww3i410, 2-amino-5-hydroxy-benzoic acid, 5-hydroxyanthranillicacid, pubchem15307, 4-amino-3-carboxyphenol, 5-hydroxyanthranillic acid | |
C1=CC(=C(C=C1O)C(=O)O)N | |
153.137 | |
153.14 | |
2-Amino-5-hydroxybenzoic acid |
∽248°C (decomposition) | |
MFCD00007870 | |
2803663 | |
HYNQTSZBTIOFKH-UHFFFAOYSA-N | |
2-amino-5-hydroxybenzoic acid | |
164592 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only