missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-fluoropyridine, 97%

Manufacturer:  Alfa Aesar™ L2001003

 View more versions of this product

Catalog No. AAL2001003



Specifications

Specifications

2-Amino-5-fluoropyridine
93°C to 97°C
21717-96-4
MFCD01861120
385938
YJTXQLYMECWULH-UHFFFAOYSA-N
5-fluoropyridin-2-amine
2737701
97%
Characteristic
1g
C5H5FN2
UN2811
2-amino 5-fluoropyridine
C1=CC(=NC=C1F)N
112.107
112.11

Chemical Identifiers

21717-96-4
112.107
YJTXQLYMECWULH-UHFFFAOYSA-N
2737701
C1=CC(=NC=C1F)N
C5H5FN2
MFCD01861120
2-amino 5-fluoropyridine
5-fluoropyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents