Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-fluoropyridine, 97%


Quantity 250mg

Manufacturer: alfa aesar™  L20010MD

 View more versions of this product

Catalog No. AAL20010MD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 112.11
Chemical Name or Material 2-Amino-5-fluoropyridine
Assay 97%
Melting Point 93° to 97°C
MDL Number MFCD01861120
CAS 21717-96-4
UN Number UN2811
Beilstein 385938
Synonym 5-Fluoro-2-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C5H5FN2
Quantity 250mg
Odor Characteristic
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Specifications

Chemical Name or Material 2-Amino-5-fluoropyridine
CAS 21717-96-4
Molecular Formula C5H5FN2
Formula Weight 112.11
MDL Number MFCD01861120
UN Number UN2811
Beilstein 385938
Synonym 5-Fluoro-2-pyridinamine
Accessories

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents