missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-fluoropyridine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC397290010

 View more versions of this product

Catalog No. AC397290010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Powder
≥94% (GC)
White to orange
94.0
2-Amino-5-fluoropyridine
21717-96-4
MFCD01861120
Glass bottle
112.11
1g
93° to 99°C
100.0
C5H5FN2
95%
Authentic

Chemical Identifiers

21717-96-4
100.0
C5H5FN2
MFCD01861120
≥94% (GC)
94.0
95%
112.11
2-Amino-5-fluoropyridine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.