missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-fluorobenzothiazole, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5102203
Description
2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2-Amino-5-fluorobenzothiazole | |
C7H5FN2S | |
1 g | |
Soluble in methanol. | |
C1=CC2=C(C=C1F)N=C(S2)N | |
168.189 | |
168.19 |
20358-07-0 | |
MFCD05664560 | |
2-amino-5-fluorobenzothiazole, 5-fluorobenzo d thiazol-2-amine, 2-benzothiazolamine, 5-fluoro, 5-fluoro-2-benzothiazolamine, 2-benzothiazolamine,5-fluoro, 5-fluorobenzothiazol-2-ylamine, pubchem21850, acmc-20a67y, 2-amino-5-fluorobenzothiozole, 5-fluoro-2-aminobenzothiazole | |
YHBIGBYIUMCLJS-UHFFFAOYSA-N | |
5-fluoro-1,3-benzothiazol-2-amine | |
821203 | |
98% |
Chemical Identifiers
20358-07-0 | |
168.189 | |
YHBIGBYIUMCLJS-UHFFFAOYSA-N | |
5-fluoro-1,3-benzothiazol-2-amine |
C7H5FN2S | |
MFCD05664560 | |
821203 | |
C1=CC2=C(C=C1F)N=C(S2)N |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only