missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-fluorobenzoic acid, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar B2145806

 View more versions of this product

Catalog No. AAB2145806



Specifications

Specifications

2-Amino-5-fluorobenzoic acid
≥98%
182°C to 184°C
2803664
5-Fluoroanthranilic acid
C1=CC(=C(C=C1F)C(=O)O)N
155.128
CHEBI:78042
446-08-2
C7H6FNO2
MFCD00055566
5g
FPQMGQZTBWIHDN-UHFFFAOYSA-N
2-amino-5-fluorobenzoic acid
101412
155.13

Chemical Identifiers

446-08-2
155.128
FPQMGQZTBWIHDN-UHFFFAOYSA-N
101412
2-amino-5-fluorobenzoic acid
C7H6FNO2
MFCD00055566
5-Fluoroanthranilic acid
CHEBI:78042
C1=CC(=C(C=C1F)C(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents