missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-fluoro-3-nitropyridine, 98%

Manufacturer:  Alfa Aesar™ H6129303

 View more versions of this product

Catalog No. AAH6129303


 

Specifications

Specifications

2-Amino-5-fluoro-3-nitropyridine
98%
MFCD04972413
1g
LDYYBZNEWDTDEE-UHFFFAOYSA-N
5-fluoro-3-nitropyridin-2-amine
21717339
Crystalline
212268-12-7
C5H4FN3O2
Orange
2-Amino-3-nitro-5-fluoropyridine
C1=C(C(=NC=C1F)N)[N+](=O)[O-]
157.104
157.1

Chemical Identifiers

212268-12-7
157.104
LDYYBZNEWDTDEE-UHFFFAOYSA-N
21717339
C5H4FN3O2
MFCD04972413
2-Amino-3-nitro-5-fluoropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents