Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-ethylthio-1,3,4-thiadiazole, 96%

$61.47 - $221.50

Chemical Identifiers

Chemical Name or Material 2-Amino-5-ethylthio-1,3,4-thiadiazole
CAS 25660-70-2
Molecular Formula C4H7N3S2
Formula Weight 161.25
MDL Number MFCD00040401
Synonym 5-(Ethylthio)-1,3,4-thiadiazole-2-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2562406

  SDS
alfa aesar™
B2562406
5g Each for $61.47

AAB2562414

  SDS
alfa aesar™
B2562414
25g Each for $221.50
Description & Specifications

Specifications

Formula Weight 161.25
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 130° to 133°C
CAS 25660-70-2
Chemical Name or Material 2-Amino-5-ethylthio-1,3,4-thiadiazole
TSCA TSCA
Quantity 5g
Assay 96%
MDL Number MFCD00040401
Molecular Formula C4H7N3S2
Health Hazard 1 Warning
Synonym 5-(Ethylthio)-1,3,4-thiadiazole-2-amine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.