Please login to your online account to display your discounted pricing

2-Amino-5-ethyl-1,3,4-thiadiazole, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  188480250

 View more versions of this product

Catalog No. AC188480250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color Light beige
Quantity 25g
Chemical Name or Material 2-Amino-5-ethyl-1, 3, 4-thiadiazole
Name Note 98+%
CAS 14068-53-2
CAS Min % 98.0
CAS Max % 100.0
Physical Form Light beige crystalline powder
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C4H7N3S
Formula Weight 129.19g/mol
MDL Number MFCD00003111
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number XI3095000
EINECS Number 237-921-4
TSCA TSCA

Specifications

Chemical Name or Material 2-Amino-5-ethyl-1, 3, 4-thiadiazole
Name Note 98+%
CAS 14068-53-2
CAS Min % 98.0
CAS Max % 100.0
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C4H7N3S
Formula Weight 129.19g/mol
MDL Number MFCD00003111
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents