Please login to your online account to display your discounted pricing

2-Amino-5-ethyl-1,3,4-thiadiazole, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-5-ethyl-1, 3, 4-thiadiazole
Name Note 98+%
CAS 14068-53-2
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H7N3S
Formula Weight 129.19
MDL Number MFCD00003111
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC188480050

  SDS
thermo fisher scientific
188480050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC188480250

  SDS
thermo fisher scientific
188480250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 129.19
Quantity 5g
Color Light beige
EINECS Number 237-921-4
TSCA TSCA
RTECS Number XI3095000
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 2-Amino-5-ethyl-1, 3, 4-thiadiazole
Molecular Formula C4H7N3S
CAS 14068-53-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98+%
Assay Percent Range 98+%
MDL Number MFCD00003111
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.