missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-ethyl-1,3,4-thiadiazole, 97%

$35.86 - $35.86

Chemical Identifiers

CAS 14068-53-2
Molecular Formula C4H7N3S
Formula Weight 129.18
MDL Number MFCD00003111
Beilstein 113614
Chemical Name or Material 2-Amino-5-ethyl-1,3,4-thiadiazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0691506 SDS Alfa Aesar™
L0691506
5g Each for $35.86
Specifications Description & Specifications

Chemical Identifiers

14068-53-2
129.18
113614
C4H7N3S
MFCD00003111
2-Amino-5-ethyl-1,3,4-thiadiazole

Specifications

14068-53-2
5g
129.18
113614
97%
198° to 202°C
C4H7N3S
MFCD00003111
2-Amino-5-ethyl-1,3,4-thiadiazole

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XI3095000

TSCA : TSCA