missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-(diethylamino)toluene Monohydrochloride 98.0+%, TCI America™

$61.11 - $520.98

Chemical Identifiers

CAS 2051-79-8
Molecular Formula C11H19ClN2
Molecular Weight (g/mol) 214.737
MDL Number MFCD00035466
InChI Key MPLZNPZPPXERDA-UHFFFAOYSA-N
Synonym 2-amino-5-diethylamino toluene monohydrochloride, 4-n,n-diethyl-2-methyl-p-phenylenediamine monohydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, monohydrochloride, unii-39mih70e0i, n4,n4-diethyl-2-methylbenzene-1,4-diamine hydrochloride, 2-amino-5-diethylaminotoluene monohydrochloride, 4-amino-3-methylphenyl diethylamine, chloride, 4-diethylamino-o-toluidine hydrochloride, n,n-diethyltoluene-2,5-diamine hydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, hydrochloride 1:1
PubChem CID 108623
IUPAC Name 4-N,4-N-diethyl-2-methylbenzene-1,4-diamine;hydrochloride
SMILES CCN(CC)C1=CC(=C(C=C1)N)C.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A028925G View Documents TCI America
A028925G
25g Each for $61.11
 
A0289500G View Documents TCI America
A0289500G
500g Each for $520.98
 
Specifications

Chemical Identifiers

2051-79-8
214.737
MPLZNPZPPXERDA-UHFFFAOYSA-N
108623
CCN(CC)C1=CC(=C(C=C1)N)C.Cl
C11H19ClN2
MFCD00035466
2-amino-5-diethylamino toluene monohydrochloride, 4-n,n-diethyl-2-methyl-p-phenylenediamine monohydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, monohydrochloride, unii-39mih70e0i, n4,n4-diethyl-2-methylbenzene-1,4-diamine hydrochloride, 2-amino-5-diethylaminotoluene monohydrochloride, 4-amino-3-methylphenyl diethylamine, chloride, 4-diethylamino-o-toluidine hydrochloride, n,n-diethyltoluene-2,5-diamine hydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, hydrochloride 1:1
4-N,4-N-diethyl-2-methylbenzene-1,4-diamine;hydrochloride

Specifications

White
2051-79-8
MFCD00035466
2-amino-5-diethylamino toluene monohydrochloride, 4-n,n-diethyl-2-methyl-p-phenylenediamine monohydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, monohydrochloride, unii-39mih70e0i, n4,n4-diethyl-2-methylbenzene-1,4-diamine hydrochloride, 2-amino-5-diethylaminotoluene monohydrochloride, 4-amino-3-methylphenyl diethylamine, chloride, 4-diethylamino-o-toluidine hydrochloride, n,n-diethyltoluene-2,5-diamine hydrochloride, 1,4-benzenediamine, n4,n4-diethyl-2-methyl-, hydrochloride 1:1
CCN(CC)C1=CC(=C(C=C1)N)C.Cl
214.737
214.74
≥98.0% (HPLC,T)
25g
C11H19ClN2
2811
MPLZNPZPPXERDA-UHFFFAOYSA-N
4-N,4-N-diethyl-2-methylbenzene-1,4-diamine;hydrochloride
108623
Crystal-Powder at 20°C
2-Amino-5-(diethylamino)toluene Monohydrochloride
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0279

RTECSNumber : SS9560000

TSCA : Yes