missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals
$96.74 - $323.84
Chemical Identifiers
CAS | 57235-50-4 |
---|---|
Molecular Formula | C5H7N3S |
Molecular Weight (g/mol) | 141.19 |
MDL Number | MFCD00051649 |
InChI Key | AVLUMBXGKFNNAS-UHFFFAOYSA-N |
Synonym | 2-amino-5-cyclopropyl-1,3,4-thiadiazole, 5-cyclopropyl 1,3,4 thiadiazol-2-ylamine, 5-cyclopropyl-1,3,4-thiadiazole-2-ylamine, acmc-20ao4n, enamine_005503, 5-cyclopropyl-2-amino-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine,5-cyclopropyl, 5-cyclopropyl-1,3,4 thiadiazol-2-ylamine |
PubChem CID | 717021 |
IUPAC Name | 5-cyclopropyl-1,3,4-thiadiazol-2-amine |
SMILES | NC1=NN=C(S1)C1CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0577803
|
Thermo Scientific Chemicals
L0577803 |
1 g |
N/A
|
|
|||||
AAL0577806
|
Thermo Scientific Chemicals
L0577806 |
5 g |
N/A
|
|
|||||
Description
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.
Solubility
Soluble in Methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at -20°C.
Chemical Identifiers
57235-50-4 | |
141.19 | |
AVLUMBXGKFNNAS-UHFFFAOYSA-N | |
717021 | |
NC1=NN=C(S1)C1CC1 |
C5H7N3S | |
MFCD00051649 | |
2-amino-5-cyclopropyl-1,3,4-thiadiazole, 5-cyclopropyl 1,3,4 thiadiazol-2-ylamine, 5-cyclopropyl-1,3,4-thiadiazole-2-ylamine, acmc-20ao4n, enamine_005503, 5-cyclopropyl-2-amino-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine,5-cyclopropyl, 5-cyclopropyl-1,3,4 thiadiazol-2-ylamine | |
5-cyclopropyl-1,3,4-thiadiazol-2-amine |
Specifications
57235-50-4 | |
C5H7N3S | |
1 g | |
2-amino-5-cyclopropyl-1,3,4-thiadiazole, 5-cyclopropyl 1,3,4 thiadiazol-2-ylamine, 5-cyclopropyl-1,3,4-thiadiazole-2-ylamine, acmc-20ao4n, enamine_005503, 5-cyclopropyl-2-amino-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine,5-cyclopropyl, 5-cyclopropyl-1,3,4 thiadiazol-2-ylamine | |
AVLUMBXGKFNNAS-UHFFFAOYSA-N | |
5-cyclopropyl-1,3,4-thiadiazol-2-amine | |
717021 | |
98% |
212°C to 216°C | |
MFCD00051649 | |
124356 | |
Soluble in Methanol. Insoluble in water. | |
NC1=NN=C(S1)C1CC1 | |
141.19 | |
141.2 | |
2-Amino-5-cyclopropyl-1,3,4-thiadiazole |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only