Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-chloropyrimidine, 97%

$39.26 - $390.97

Chemical Identifiers

Chemical Name or Material 2-Amino-5-chloropyrimidine
CAS 5428-89-7
Molecular Formula C4H4ClN3
Formula Weight 129.55
MDL Number MFCD00014604
Beilstein 110914
Synonym 5-Chloro-2-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1924803

  SDS
alfa aesar™
L1924803
1g Each for $97.89

AAL1924806

  SDS
alfa aesar™
L1924806
5g Each for $390.97

AAL19248MD

  SDS
alfa aesar™
L19248MD
250mg Each for $39.26
Description & Specifications

Specifications

Formula Weight 129.55
Chemical Name or Material 2-Amino-5-chloropyrimidine
Assay 97%
MDL Number MFCD00014604
CAS 5428-89-7
TSCA TSCA
Quantity 1g
Molecular Formula C4H4ClN3
Beilstein 110914
Synonym 5-Chloro-2-pyrimidinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UV6329500
Melting Point 235°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.