missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyridine, 98%

CAS: 1072-98-6 | C5H5ClN2 | 128.559 g/mol

Supplier:  Thermo Scientific Chemicals A1206322

 View more versions of this product

Catalog No. AAA1206322


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-5-chloropyridine
135°C to 138°C
160°C (320°F)
MFCD00006324
108735
MAXBVGJEFDMHNV-UHFFFAOYSA-N
5-chloropyridin-2-amine
66174
98%
1072-98-6
127°C to 129°C (21 mmHg)
C5H5ClN2
100 g
2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
C1=CC(=NC=C1Cl)N
128.559
128.56

Chemical Identifiers

1072-98-6
128.559
MAXBVGJEFDMHNV-UHFFFAOYSA-N
66174
C1=CC(=NC=C1Cl)N
C5H5ClN2
MFCD00006324
2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
5-chloropyridin-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 214-020-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only