missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-chloropyridine 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 103410250

 View more versions of this product

Catalog No. AC103410250


 

Specifications Description & Specifications

Specifications

≥97.5% (HPLC)
Crystalline Powder
Beige to beige-brown
100.0
Plastic bottle
97.5
98%
MFCD00006324
C5H5ClN2
128.56
25g
134° to 138°C
22, V,8, 451
Cl(C5H3N)NH2
98%
1072-98-6
2-Amino-5-chloropyridine
Authentic

Chemical Identifiers

1072-98-6
100.0
C5H5ClN2
128.56
2-Amino-5-chloropyridine, 98%
97.5
98%
Cl(C5H3N)NH2
MFCD00006324
≥97.5% (HPLC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 214-020-4