missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-chloropyridine, 98%, ACROS Organics™
$69.49 - $69.49
Chemical Identifiers
CAS | 1072-98-6 |
---|---|
Molecular Formula | C5H5ClN2 |
Molecular Weight (g/mol) | 128.56 |
MDL Number | MFCD00006324 |
InChI Key | MAXBVGJEFDMHNV-UHFFFAOYSA-N |
Synonym | 2-amino-5-chloro pyridine, 2-amino-5-chloropyridine, 2-amino5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-aminopyridine, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro |
PubChem CID | 66174 |
IUPAC Name | 5-chloropyridin-2-amine |
SMILES | C1=CC(=NC=C1Cl)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC103410250
|
Thermo Fisher Scientific
103410250 |
25g | Plastic bottle |
Each for $69.49
|
|
AC103411000
|
Thermo Fisher Scientific
103411000 |
100g | Plastic bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC103415000
|
Thermo Fisher Scientific
103415000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1072-98-6 | |
128.56 | |
MAXBVGJEFDMHNV-UHFFFAOYSA-N | |
66174 | |
C1=CC(=NC=C1Cl)N |
C5H5ClN2 | |
MFCD00006324 | |
2-amino-5-chloro pyridine, 2-amino-5-chloropyridine, 2-amino5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-aminopyridine, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro | |
5-chloropyridin-2-amine |
Specifications
160°C | |
Plastic bottle | |
Beige to Beige-Brown | |
25g | |
97.5 | |
97.5% min. (HPLC) | |
Cl(C5H3N)NH2 | |
22, V,8, 451 | |
Solubility in water: 1g/L water (20°C) | |
C1=CC(=NC=C1Cl)N | |
128.56 | |
128.56 | |
98% |
Authentic | |
127.0°C to 128.0°C (11.0 mmHg) | |
134.0°C to 138.0°C | |
1072-98-6 | |
100.0 | |
C5H5ClN2 | |
MFCD00006324 | |
2-amino-5-chloro pyridine, 2-amino-5-chloropyridine, 2-amino5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-aminopyridine, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro | |
MAXBVGJEFDMHNV-UHFFFAOYSA-N | |
5-chloropyridin-2-amine | |
66174 | |
Crystalline Powder | |
2-Amino-5-chloropyridine, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 214-020-4