missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-chloropyridine, 98%

Manufacturer:  Alfa Aesar™ A1206336

 View more versions of this product

Catalog No. AAA1206336



Specifications

Specifications

2-Amino-5-chloropyridine
127°C to 129°C (21mmHg)
500g
C5H5ClN2
108735
MAXBVGJEFDMHNV-UHFFFAOYSA-N
5-chloropyridin-2-amine
66174
98%
160°C (320°F)
135°C to 138°C
1072-98-6
MFCD00006324
2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
C1=CC(=NC=C1Cl)N
128.559
128.56

Chemical Identifiers

1072-98-6
128.559
MAXBVGJEFDMHNV-UHFFFAOYSA-N
66174
C1=CC(=NC=C1Cl)N
C5H5ClN2
MFCD00006324
2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
5-chloropyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

SDS Documents