Please login to your online account to display your discounted pricing

2-Amino-5-chlorophenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-5-chlorophenylboronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C12H17BClNO2
Formula Weight 253.54
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444470010

 
thermo fisher scientific
AC444470010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444470050

 
thermo fisher scientific
AC444470050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.
Formula Weight 253.54
Quantity 1g
Chemical Name or Material 2-Amino-5-chlorophenylboronic acid pinacol ester
Molecular Formula C12H17BClNO2
Health Hazard 1 Warning
Assay Percent Range 97%
CAS 1073371-77-3
Safety and Handling

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation.