missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-chlorobenzophenone, 98+%

Manufacturer:  Alfa Aesar™ A1572036

 View more versions of this product

Catalog No. AAA1572036



Description

Description

Applications
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2-Amino-5-chlorobenzophenone
96°C to 99°C
719-59-5
MFCD00007839
2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Cl)N
231.679
231.68
211°C (412°F)
500g
C13H10ClNO
475640
ZUWXHHBROGLWNH-UHFFFAOYSA-N
(2-amino-5-chlorophenyl)-phenylmethanone
12870
≥98%

Chemical Identifiers

719-59-5
231.679
ZUWXHHBROGLWNH-UHFFFAOYSA-N
12870
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)Cl)N
C13H10ClNO
MFCD00007839
2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone
(2-amino-5-chlorophenyl)-phenylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : PC4933500

TSCA : TSCA

Documents