Please login to your online account to display your discounted pricing

2-Amino-5-chlorobenzoic Acid 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  103331000

 View more versions of this product

Catalog No. AC103331000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 171.58
Quantity 100g
Beilstein 14, 365
EINECS Number 211-230-8
TSCA TSCA
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Amino-5-chlorobenzoic acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7 H6 Cl N O2
Linear Formula H2NC6H3(Cl)CO2H
MDL Number MFCD00007838
CAS 635-21-2
CAS Min % 97.5
Packaging Glass bottle
Synonym 5-Chloroanthranilic acid

Specifications

Chemical Name or Material 2-Amino-5-chlorobenzoic acid
Name Note 98%
CAS 635-21-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7 H6 Cl N O2
Linear Formula H2NC6H3(Cl)CO2H
Formula Weight 171.58
MDL Number MFCD00007838
Synonym 5-Chloroanthranilic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents