Please login to your online account to display your discounted pricing

2-Amino-5-chlorobenzoic Acid 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-5-chlorobenzoic acid
Name Note 98%
CAS 635-21-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7 H6 Cl N O2
Linear Formula H2NC6H3(Cl)CO2H
Formula Weight 171.58
MDL Number MFCD00007838
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC103331000

  SDS
thermo fisher scientific
103331000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.58
Quantity 100g
Beilstein 14, 365
EINECS Number 211-230-8
TSCA TSCA
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Amino-5-chlorobenzoic acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7 H6 Cl N O2
Linear Formula H2NC6H3(Cl)CO2H
MDL Number MFCD00007838
CAS 635-21-2
CAS Min % 97.5
Packaging Glass bottle
Synonym 5-Chloroanthranilic acid
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.