Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-chloro-3-nitropyridine, 98+%

$227.81 - $631.11

Chemical Identifiers

Chemical Name or Material 2-Amino-5-chloro-3-nitropyridine
CAS 5409-39-2
Molecular Formula C5H4ClN3O2
Formula Weight 173.56
MDL Number MFCD00092011
Beilstein 383850
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2248414

  SDS
alfa aesar™
B2248414
25g Each for $227.81

AAB2248422

  SDS
alfa aesar™
B2248422
100g Each for $631.11
Description & Specifications

Specifications

Formula Weight 173.56
Chemical Name or Material 2-Amino-5-chloro-3-nitropyridine
Molecular Formula C5H4ClN3O2
Beilstein 383850
Health Hazard 1 Warning
Melting Point 190° to 195°C
Quantity 25g
Assay ≥98%
MDL Number MFCD00092011
CAS 5409-39-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.