Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-chloro-3-fluoropyridine, 98%

$73.26 - $678.82

Chemical Identifiers

Chemical Name or Material 2-Amino-5-chloro-3-fluoropyridine
CAS 246847-98-3
Molecular Formula C5H4ClFN2
Formula Weight 146.55
MDL Number MFCD09878434
Synonym 5-Chloro-3-fluoro-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3013503

  SDS
alfa aesar™
H3013503
1g Each for $204.73

AAH30135MD

  SDS
alfa aesar™
H30135MD
250mg Each for $73.26

AAH3013506

  SDS
alfa aesar™
H3013506
5g Each for $678.82
Description & Specifications

Specifications

Formula Weight 146.55
Chemical Name or Material 2-Amino-5-chloro-3-fluoropyridine
Melting Point 95° to 97°C
Quantity 1g
Assay 98%
Molecular Formula C5H4ClFN2
MDL Number MFCD09878434
CAS 246847-98-3
Synonym 5-Chloro-3-fluoro-2-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.