missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-chloro-2'-fluorobenzophenone, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 296370250

 View more versions of this product

Catalog No. AC296370250


 

Specifications Description & Specifications

Specifications

≥98.5%
249.67
Yellow
100.0
99%
C13H9ClFNO
MFCD00038381
Authentic
Fine Powder
25g
93° to 97°C
2-Amino-5-chloro-2'-fluorobenzophenone
98.5
784-38-3
99%

Chemical Identifiers

784-38-3
100.0
C13H9ClFNO
MFCD00038381
≥98.5%
98.5
99%
249.67
2-Amino-5-chloro-2'-fluorobenzophenone, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 212-316-8

TSCA : TSCA