Please login to your online account to display your discounted pricing

2-Amino-5-chloro-2'-fluorobenzophenone, 99%, Acros Organics

$164.55 - $164.55

Chemical Identifiers

Chemical Name or Material 2-Amino-5-chloro-2'-fluorobenzophenone
Name Note 99%
CAS 784-38-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H9ClFNO
Formula Weight 249.67
MDL Number MFCD00038381
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC296370250

  SDS
thermo fisher scientific
296370250
Each for $164.55
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Powder
Formula Weight 249.67
Quantity 25g
Color Yellow
TSCA TSCA
EINECS Number 212-316-8
Melting Point 93° to 97°C
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Amino-5-chloro-2'-fluorobenzophenone
Assay Percent Range 99%
CAS Min % 98.5
Molecular Formula C13H9ClFNO
CAS 784-38-3
MDL Number MFCD00038381
Name Note 99%
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.