missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromobenzothiazole, 97%

Manufacturer:  Alfa Aesar™ H5527003

 View more versions of this product

Catalog No. AAH5527003



Description

Description

Applications
2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Amino-5-bromobenzothiazole
194°C to 198°C
20358-03-6
MFCD00577806
ZPUJTWBWSOOMRP-UHFFFAOYSA-N
5-bromo-1,3-benzothiazol-2-amine
13775785
Solid
White to Brown
1g
C7H5BrN2S
2-amino-5-bromo benzothiazole
C1=CC2=C(C=C1Br)N=C(S2)N
229.095
229.1
97%

Chemical Identifiers

20358-03-6
229.095
ZPUJTWBWSOOMRP-UHFFFAOYSA-N
13775785
C1=CC2=C(C=C1Br)N=C(S2)N
C7H5BrN2S
MFCD00577806
2-amino-5-bromo benzothiazole
5-bromo-1,3-benzothiazol-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents