missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromobenzothiazole, 97%

Manufacturer:  Alfa Aesar™ H5527014

 View more versions of this product

Catalog No. AAH5527014



Description

Description

Applications
2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Amino-5-bromobenzothiazole
194°C to 198°C
20358-03-6
MFCD00577806
Soluble in water.
C1=CC2=C(C=C1Br)N=C(S2)N
229.095
229.1
97%
White to Brown
25g
C7H5BrN2S
2-amino-5-bromo benzothiazole
ZPUJTWBWSOOMRP-UHFFFAOYSA-N
5-bromo-1,3-benzothiazol-2-amine
13775785
Solid

Chemical Identifiers

20358-03-6
229.095
ZPUJTWBWSOOMRP-UHFFFAOYSA-N
13775785
C1=CC2=C(C=C1Br)N=C(S2)N
C7H5BrN2S
MFCD00577806
2-amino-5-bromo benzothiazole
5-bromo-1,3-benzothiazol-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents