Learn More
2-Amino-5-bromobenzonitrile, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 350250050
Specifications
2-Amino-5-bromobenzonitrile | |
96.0 | |
93°C to 98°C | |
Authentic | |
Glass bottle | |
MFCD00158946 | |
4-bromo-2-cyanoaniline, 2-amino-5-brombenzonitrile, 2-amio-5-bromobenzonitrile, 5-bromoanthranilonitrile, 2-amino-5-bromobenzenecarbonitrile, 2-amino-5-bromo-benzonitrile, benzonitrile, 2-amino-5-bromo, 2-bromo-5-cyanoaniline, pubchem4602, 2-cyano-4-bromoaniline | |
NC1=CC=C(Br)C=C1C#N | |
197.04 | |
197.03 | |
Crystalline Powder |
39263-32-6 | |
100.0 | |
White to Brown | |
96% min. (GC) | |
C7H5BrN2 | |
5 g | |
OATYCBHROMXWJO-UHFFFAOYSA-N | |
2-amino-5-bromobenzonitrile | |
429740 | |
97% |
Chemical Identifiers
39263-32-6 | |
197.04 | |
OATYCBHROMXWJO-UHFFFAOYSA-N | |
429740 | |
NC1=CC=C(Br)C=C1C#N |
C7H5BrN2 | |
MFCD00158946 | |
4-bromo-2-cyanoaniline, 2-amino-5-brombenzonitrile, 2-amio-5-bromobenzonitrile, 5-bromoanthranilonitrile, 2-amino-5-bromobenzenecarbonitrile, 2-amino-5-bromo-benzonitrile, benzonitrile, 2-amino-5-bromo, 2-bromo-5-cyanoaniline, pubchem4602, 2-cyano-4-bromoaniline | |
2-amino-5-bromobenzonitrile |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 254-387-8
TSCA : TSCA
RUO – Research Use Only