missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromobenzonitrile, 96%

Manufacturer:  Alfa Aesar™ H5521703

 View more versions of this product

Catalog No. AAH5521703



Description

Description

Applications
2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

2-Amino-5-bromobenzonitrile
1g
C7H5BrN2
UN3439
Soluble in methanol.
C1=CC(=C(C=C1Br)C#N)N
197.035
197.03
96°C to 100°C
39263-32-6
MFCD00158946
2-amino-5-brombenzonitrile
OATYCBHROMXWJO-UHFFFAOYSA-N
2-amino-5-bromobenzonitrile
429740
96%

Chemical Identifiers

39263-32-6
197.035
OATYCBHROMXWJO-UHFFFAOYSA-N
429740
C1=CC(=C(C=C1Br)C#N)N
C7H5BrN2
MFCD00158946
2-amino-5-brombenzonitrile
2-amino-5-bromobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 254-387-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents