missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1191506

 View more versions of this product

Catalog No. AAA1191506



Description

Description

Applications
It is used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. Also used in the determination of cobalt, copper, nickel, and zinc.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

2-Amino-5-bromobenzoic acid
98%
C7H6BrNO2
5g
14,1405
CUKXRHLWPSBCTI-UHFFFAOYSA-N
2-amino-5-bromobenzoic acid
79858
5794-88-7
~215°C (decomposition)
MFCD00007823
639028
5-Bromoanthranilic acid
C1=CC(=C(C=C1Br)C(=O)O)N
216.034
216.03

Chemical Identifiers

5794-88-7
216.034
CUKXRHLWPSBCTI-UHFFFAOYSA-N
79858
C1=CC(=C(C=C1Br)C(=O)O)N
C7H6BrNO2
MFCD00007823
5-Bromoanthranilic acid
2-amino-5-bromobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : CB2557670

SDS Documents