Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromobenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2-Amino-5-bromobenzaldehyde
CAS 29124-57-0
Formula Weight 200
MDL Number MFCD10696879
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3349103

  SDS
alfa aesar™
H3349103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH33491MD

  SDS
alfa aesar™
H33491MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 75° to 77°C
Formula Weight 200
Sensitivity Air Sensitive
Assay 97%
Health Hazard 1 Warning
Chemical Name or Material 2-Amino-5-bromobenzaldehyde
Color Yellow
Quantity 1g
CAS 29124-57-0
MDL Number MFCD10696879
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
Participate in synthesis and structure activity of novel camptothecin analogs.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.