missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine, 95%

$29.45 - $345.43

Chemical Identifiers

CAS 935534-47-7
Molecular Formula C5H3BrF3N3
Molecular Weight (g/mol) 241.999
MDL Number MFCD09261254
InChI Key FAMGPURZLOTOKD-UHFFFAOYSA-N
Synonym 5-bromo-4-trifluoromethyl pyrimidin-2-amine, 5-bromo-4-trifluoromethyl-pyrimidin-2-ylamine, 5-bromo-4-trifluoromethyl-2-pyrimidinamine, ksc672m4h, 2-amino-5-bromo-4-trifluoromethylpyrimidine, 5-bromo-4-trifluoromethyl-2-pyrimidineamin, 5-bromo-4-trifluoromethyl-2-pyrimidylamine, 2-amino-5-bromo-4-trifluoromethyl pyrimidine, 2-pyrimidinamine, 5-bromo-4-trifluoromethyl, 5-bromo-4-trifluoromethyl pyrimidine-2-ylamine
PubChem CID 45382152
IUPAC Name 5-bromo-4-(trifluoromethyl)pyrimidin-2-amine
SMILES C1=C(C(=NC(=N1)N)C(F)(F)F)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64470MD View Documents Alfa Aesar™
H64470MD
250mg Each for $29.45
 
AAH6447003 View Documents Alfa Aesar™
H6447003
1g Each for $86.99
 
AAH6447006 missing translation for 'viewDocuments' Alfa Aesar™
H6447006
5g Each for $345.43
 
Specifications

Chemical Identifiers

935534-47-7
241.999
FAMGPURZLOTOKD-UHFFFAOYSA-N
45382152
C1=C(C(=NC(=N1)N)C(F)(F)F)Br
C5H3BrF3N3
MFCD09261254
5-bromo-4-trifluoromethyl pyrimidin-2-amine, 5-bromo-4-trifluoromethyl-pyrimidin-2-ylamine, 5-bromo-4-trifluoromethyl-2-pyrimidinamine, ksc672m4h, 2-amino-5-bromo-4-trifluoromethylpyrimidine, 5-bromo-4-trifluoromethyl-2-pyrimidineamin, 5-bromo-4-trifluoromethyl-2-pyrimidylamine, 2-amino-5-bromo-4-trifluoromethyl pyrimidine, 2-pyrimidinamine, 5-bromo-4-trifluoromethyl, 5-bromo-4-trifluoromethyl pyrimidine-2-ylamine
5-bromo-4-(trifluoromethyl)pyrimidin-2-amine

Specifications

250mg
C5H3BrF3N3
5-bromo-4-trifluoromethyl pyrimidin-2-amine, 5-bromo-4-trifluoromethyl-pyrimidin-2-ylamine, 5-bromo-4-trifluoromethyl-2-pyrimidinamine, ksc672m4h, 2-amino-5-bromo-4-trifluoromethylpyrimidine, 5-bromo-4-trifluoromethyl-2-pyrimidineamin, 5-bromo-4-trifluoromethyl-2-pyrimidylamine, 2-amino-5-bromo-4-trifluoromethyl pyrimidine, 2-pyrimidinamine, 5-bromo-4-trifluoromethyl, 5-bromo-4-trifluoromethyl pyrimidine-2-ylamine
C1=C(C(=NC(=N1)N)C(F)(F)F)Br
241.999
242
95%
935534-47-7
MFCD09261254
FAMGPURZLOTOKD-UHFFFAOYSA-N
5-bromo-4-(trifluoromethyl)pyrimidin-2-amine
45382152
Solid
2-Amino-5-bromo-4-(trifluoromethyl)pyrimidine