missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-4-methylpyrimidine, 96%

Manufacturer:  Alfa Aesar™ H5418614

 View more versions of this product

Catalog No. AAH5418614



Specifications

Specifications

2-Amino-5-bromo-4-methylpyrimidine
25g
C5H6BrN3
2-amino-4-methyl-5-bromopyrimidine
CC1=NC(=NC=C1Br)N
188.028
188.03
192°C to 194°C
17321-93-6
MFCD00094469
LPQVTZJEIXYDQA-UHFFFAOYSA-N
5-bromo-4-methylpyrimidin-2-amine
18616196
96%

Chemical Identifiers

17321-93-6
188.028
LPQVTZJEIXYDQA-UHFFFAOYSA-N
18616196
CC1=NC(=NC=C1Br)N
C5H6BrN3
MFCD00094469
2-amino-4-methyl-5-bromopyrimidine
5-bromo-4-methylpyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents