missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-4-methylpyridine, 97%

Manufacturer:  Alfa Aesar™ 4462306

 View more versions of this product

Catalog No. AA4462306



Specifications

Specifications

2-Amino-5-bromo-4-methylpyridine
148°C to 151°C
98198-48-2
MFCD03427660
Soluble in methanol.
CC1=CC(=NC=C1Br)N
187.04
187.04
97%
Cream
5g
C6H7BrN2
2-amino-4-methyl-5-bromopyridine
JDNCMHOKWINDKI-UHFFFAOYSA-N
5-bromo-4-methylpyridin-2-amine
817695
Powder

Chemical Identifiers

98198-48-2
187.04
JDNCMHOKWINDKI-UHFFFAOYSA-N
817695
CC1=CC(=NC=C1Br)N
C6H7BrN2
MFCD03427660
2-amino-4-methyl-5-bromopyridine
5-bromo-4-methylpyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents