Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-5-bromo-4-hydroxy-6-methylpyrimidine, 99%

$33.72 - $317.09

Chemical Identifiers

Chemical Name or Material 2-Amino-5-bromo-4-hydroxy-6-methylpyrimidine
CAS 6307-35-3
Molecular Formula C5H6BrN3O
Formula Weight 204.03
MDL Number MFCD00006092
Beilstein 4959
Synonym 2-Amino-5-bromo-6-methyl-4-pyrimidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2080706

 
alfa aesar™
B2080706
5g Each for $33.72

AAB2080714

 
alfa aesar™
B2080714
25g Each for $96.51

AAB2080722

 
alfa aesar™
B2080722
100g Each for $317.09
Description & Specifications

Specifications

Formula Weight 204.03
Chemical Name or Material 2-Amino-5-bromo-4-hydroxy-6-methylpyrimidine
Assay 99%
Melting Point ≈240°C (dec.)
Molecular Formula C5H6BrN3O
Quantity 5g
MDL Number MFCD00006092
CAS 6307-35-3
Beilstein 4959
Synonym 2-Amino-5-bromo-6-methyl-4-pyrimidinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.